Skip to Content
Merck
Fuchsin (C.I. 42510) for microscopy Certistain®

Fuchsin (C.I. 42510)

Fuchsin (C.I. 42510) for microscopy Certistain®
Synonym(s):
Fuchsin (C.I. 42510)
Empirical Formula (Hill Notation):
C20H20ClN3
CAS Number:
Molecular Weight:
337.85

Products

Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing