Skip to Content
Merck
Chloroform-D1 deuteration degree min. 99.95% for NMR spectroscopy MagniSolv™

Chloroform-D1

Chloroform-D1 deuteration degree min. 99.95% for NMR spectroscopy MagniSolv™
Synonym(s):
Chloroform-D1
Empirical Formula (Hill Notation):
CCl3D
CAS Number:
Molecular Weight:
120.38

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Isotope
Technique