Benz[a]anthracene solution
Synonym(s):
Benzo[a]anthracene solution
Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
Products
Brand
Formula Weight
Gene Alias
Product Line
Format
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing