
Acetoxyvalerenic acid

Synonym(s):
Acetylvalerenolic acid
Empirical Formula (Hill Notation):
C17H24O4
CAS Number:
Molecular Weight:
292.37
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1