Skip to Content
Merck
4-Methylquinoline ≥99%

4-Methylquinoline

4-Methylquinoline ≥99%
Synonym(s):
Lepidine, 4-Methylquinoline
Empirical Formula (Hill Notation):
C10H9N
CAS Number:
Molecular Weight:
143.19
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Showing 1-1 of 1
Compare
Product Number
Description
Pricing