Skip to Content
Merck
4-Allylanisole ≥98%, FCC

4-Allylanisole

4-Allylanisole ≥98%, FCC
Synonym(s):
p-Allylphenyl methyl ether, p-Methoxyallylbenzene, Chavicol methyl ether, Estragole, Isoanethole, Methyl chavicol
Linear Formula:
H2C=CHCH2C6H4OCH3
CAS Number:
Molecular Weight:
148.20
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing