
4,4′-Thiobis(2-tert-butyl-5-methylphenol)

Synonym(s):
4,4′-Thiobis(2-tert-butyl-5-methylphenol)
Linear Formula:
(2-(CH3)-4-(OH)-5-((CH3)3C)C6H2)2S
CAS Number:
Molecular Weight:
358.54
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing