
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate

Synonym(s):
MBTH hydrochloride hydrate
Empirical Formula (Hill Notation):
C8H9N3S · HCl · xH2O
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing