Skip to Content
Merck
2-Methylhexanoic acid ≥99%, FG

2-Methylhexanoic acid

2-Methylhexanoic acid ≥99%, FG
Synonym(s):
2-Methylcaproic acid
Linear Formula:
CH3(CH2)3CH(CH3)COOH
CAS Number:
Molecular Weight:
130.18
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing