2-Chloro-3,5-dinitropyridine
Synonym(s):
2-Chloro-3,5-dinitropyridine
Empirical Formula (Hill Notation):
C5H2ClN3O4
CAS Number:
Molecular Weight:
203.54
Products
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing