Skip to Content
Merck
2,4-Hexadienoic acid ≥99%, FCC

2,4-Hexadienoic acid

2,4-Hexadienoic acid ≥99%, FCC
Synonym(s):
Sorbic acid, α-trans-γ-trans-Sorbic acid, (E,E)-1,3-Pentadiene-1-carboxylic acid, 2,4-Hexadienoic acid, 2E,4E-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, C6:2n-2,4, NSC 35405, NSC 49103, NSC 50268
Linear Formula:
CH3CH=CHCH=CHCOOH
CAS Number:
Molecular Weight:
112.13
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Application
Markush Class
Markush Group
Organoleptic Class
Showing 1-1 of 1
Compare
Product Number
Description
Pricing