Skip to Content
Merck
1-Methoxy-2-propanol EMPLURA®

1-Methoxy-2-propanol

1-Methoxy-2-propanol EMPLURA®
Synonym(s):
Propylene glycol methyl ether, Propyleneglycol monomethyl ether, 1-Methoxy-2-propanol
Linear Formula:
CH3CH(OH)CH2OCH3
CAS Number:
Molecular Weight:
90.12

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing