Skip to Content
Merck
1-Boc-piperazine 97%

1-Boc-piperazine

1-Boc-piperazine 97%
Synonym(s):
tert-Butyl piperazine-1-carboxylate
Empirical Formula (Hill Notation):
C9H18N2O2
CAS Number:
Molecular Weight:
186.25

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing