1,8-Dihydroxyanthraquinone
Synonym(s):
1,8-Dihydroxyanthraquinone
Empirical Formula (Hill Notation):
C14H8O4
CAS Number:
Molecular Weight:
240.21
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing