Skip to Content
Merck
1,2-Dimethoxybenzene ≥99%, FG

1,2-Dimethoxybenzene

1,2-Dimethoxybenzene ≥99%, FG
Synonym(s):
Pyrocatechol dimethyl ether, Veratrole
Linear Formula:
C6H4(OCH3)2
CAS Number:
Molecular Weight:
138.16
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing