Product Name
N-Adipoylglycine analytical standard
Product Number
53603
Product Brand
sial
CAS Number
66060-81-9
Molecular Weight
203.19
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 95.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS