Skip to Content
Merck
All Photos(1)

Key Documents

A8652

Sigma-Aldrich

Stearyl arachidate

Synonym(s):

Arachidic acid stearyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H76O2
CAS Number:
Molecular Weight:
565.01
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥98% (GC)

form

powder

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCCCC(=O)OCCCCCCCCCCCCCCCCCC

InChI

1S/C38H76O2/c1-3-5-7-9-11-13-15-17-19-21-22-24-26-28-30-32-34-36-38(39)40-37-35-33-31-29-27-25-23-20-18-16-14-12-10-8-6-4-2/h3-37H2,1-2H3

InChI key

XPRSWAIUFMEQLF-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luciana Mercedes Pujol-Lereis et al.
Journal of insect physiology, 87, 53-62 (2016-02-13)
The remodeling of membrane composition by changes in phospholipid head groups and fatty acids (FA) degree of unsaturation has been associated with the maintenance of membrane homeostasis under stress conditions. Overall lipid levels and the composition of cuticle lipids also
Jhon J López-Perea et al.
Environmental pollution (Barking, Essex : 1987), 250, 323-330 (2019-04-20)
The chemical composition of the uropygial gland secretion of birds shows seasonal, sex and age-related variations following sex hormones fluctuations. We explore the use of the composition of the uropygial gland secretion as a non-invasive biomarker of endocrine disruption in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service