MAT047029948
Matrix Scientific
N'-(3,4-Dimethoxy-benzyl)-N,N-dimethyl-ethane-1,2-diamine
About This Item
bp
126.5-127.5/0.3Torr
density
1.0322/25
Iupac Name
N~1~-(3,4-dimethoxybenzyl)-N~2~,N~2~-dimethyl-1,2-ethanediamine
Inchi Code
1S/C13H22N2O2/c1-15(2)8-7-14-10-11-5-6-12(16-3)13(9-11)17-4/h5-6,9,14H,7-8,10H2,1-4H3
InChI key
CISCZWBNIQPITJ-UHFFFAOYSA-N
Country of Origin
Product Link
Boiling point
density
MSDS
https://www.matrixscientific.com/010980.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service