Merck
LC/MS Analysis of Drugs of Abuse on Ascentis® Express RP-Amide
LC/MS Analysis of Drugs of Abuse on Ascentis® Express RP-Amide application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express RP-Amide, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
mobile phase component
Product No.
Description
Pricing
Ammonium formate
eluent additive for LC-MS, LiChropur, ≥99.0%
standard
Product No.
Description
Pricing
Benzoylecgonine solution
100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Cocaethylene solution
1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
EDDP perchlorate
ampule of 10 mg, certified reference material, Cerilliant®
(±)-Methadone solution
100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
CONDITIONS
column
Ascentis Express RP Amide, 10 cm x 2.1 mm I.D., 2.7 μm particles (53913-U)
mobile phase
[A] 10 mM ammonium formate in water; [B] 10mM ammonium formate in acetonitrile; (75:25, A:B)
flow rate
0.2 mL/min
column temp.
35 °C
detector
ESI(+)
injection
2 μL
sample
500 ng/mL in water
Description
Analysis Note
This applic.ation demonstrates the suitability of Ascentis Express RP-Amide for the efficient separation of common drink additives used in diet cola
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany