Merck
  • Supramolecular photocatalysis by confinement--photodimerization of coumarins within cucurbit[8]urils.
Supramolecular photocatalysis by confinement--photodimerization of coumarins within cucurbit[8]urils.
Chemical communications (Cambridge, England) (2009-12-22)
Barry C Pemberton, Nilotpal Barooah, D K Srivatsava, J Sivaguru
ABSTRACT
Cucurbit[8]uril (as low as 10 mol%) acts as a supramolecular catalytic nanoreaction vessel and facilitates the photodimerization of coumarins in water leading to syn dimers. Saturation kinetics shows a sigmoidal dependence with a turnover number of 3.4 min(-1) and a Hill constant of 1.8 indicating a co-operative mechanism in the catalytic process.
Sigma-Aldrich
6-Methylcoumarin, ≥99%
Sigma-Aldrich
6-Methylcoumarin, ≥99%, FCC, FG