Merck
  • Local and remote charge-transfer-enhanced Raman scattering on one-dimensional transition-metal oxides.
Local and remote charge-transfer-enhanced Raman scattering on one-dimensional transition-metal oxides.
Chemistry, an Asian journal (2010-06-12)
Bin Dong, Yingzhou Huang, Naisen Yu, Yurui Fang, Baosheng Cao, Yuanzuo Li, Hongxing Xu, Mengtao Sun
ABSTRACT
The one-dimensional (1D) transition-metal oxide MoO(3) belt is synthesized and characterized with X-ray diffraction, scanning electron microscopy, and Raman spectroscopy. Charge-transfer-(CT) enhanced Raman scattering of 4-mercaptobenzoic acid (4-MBA) on a 1D MoO(3) belt was investigated experimentally and theoretically. The chemical enhancement of surface-enhanced Raman scattering (SERS) of 4-MBA on the MoO(3) belt by CT is in the order of 10(3). The SERS of 4-MBA was investigated theoretically by using a quantum chemical method. The remote SERS of 4-MBA along the 1D MoO(3) belt (the light excitation to one side of the MoO(3) belt, and the SERS spectrum is collected on the other side of the MoO(3) belt) is also shown experimentally, which provides potential applications of SERS. The incident polarization dependence of remote SERS spectra has also been investigated experimentally.
Sigma-Aldrich
4-Mercaptobenzoic acid, 99%
Sigma-Aldrich
4-Mercaptobenzoic acid, technical grade, 90%