Merck
  • CHROMagar Yersinia, a new chromogenic agar for screening of potentially pathogenic Yersinia enterocolitica isolates in stools.
CHROMagar Yersinia, a new chromogenic agar for screening of potentially pathogenic Yersinia enterocolitica isolates in stools.
Journal of clinical microbiology (2013-02-01)
Nicolas Renaud, Laetitia Lecci, René J Courcol, Michel Simonet, Olivier Gaillot
ABSTRACT
CHROMagar Yersinia (CAY) is a new chromogenic medium for the presumptive detection of virulent Yersinia enterocolitica in stools. Based on a comparative analysis of 1,494 consecutive stools from hospitalized patients, CAY was found to be just as sensitive as the reference medium (cefsulodin-irgasan-novobiocin agar) but was significantly more specific and had a very low false-positive rate. CAY reduces the workload (and thus costs) for stool analysis and can therefore be recommended for routine laboratory use.
Product Description
Millipore
Agar, powder, suitable for microbiology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Agar, suitable for plant cell culture
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Sigma-Aldrich
Select agar
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Noble agar
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Millipore
Agar, high purity, low ionic content, low gel strenght, suitable for microbiology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Bacteriological agar, for molecular biology