Merck
  • No role of homologous recombination in dealing with β-lapachone cytotoxicity in yeast.
No role of homologous recombination in dealing with β-lapachone cytotoxicity in yeast.
Chemical research in toxicology (2011-11-19)
Oliver Quevedo, Jonay García-Luis, Isabel Lorenzo-Castrillejo, Félix Machín
ABSTRACT
β-Lapachone (β-lap) is a promising antitumoral agent. DNA base oxidation and alkylation are among the expected damages by β-lap. Herein, we have explored the role that the homologous recombination pathway (HR), a critical DNA repair process in Saccharomyces cerevisiae, has in the cytotoxic profile of β-lap. We have further compared β-lap to the closely related compound menadione and the well-known alkylating agent methyl methanesulfonate (MMS). Surprisingly, we found that β-lap does not trigger HR, as seen for (i) the mutant sensitivity profiles, (ii) concentration-dependent arrest profiles, (iii) absence of nuclear DNA repair factories, and (iv) frequency of recombination between direct repeats.
Sigma-Aldrich
β-Lapachone, ≥98% (TLC)