Merck
  • Pro-oxidants ameliorate radiation-induced apoptosis through activation of the calcium-ERK1/2-Nrf2 pathway.
Pro-oxidants ameliorate radiation-induced apoptosis through activation of the calcium-ERK1/2-Nrf2 pathway.
Free radical biology & medicine (2011-05-03)
Nazir M Khan, Santosh K Sandur, Rahul Checker, Deepak Sharma, T B Poduval, Krishna B Sainis
ABSTRACT
There are no reports describing the ability of pro-oxidants to protect against radiation-induced apoptosis. Activation of the redox-sensitive transcription factor Nrf2 by low levels of ROS is known to protect against oxidative stress-induced cell death. In this study, hydrogen peroxide, diethylmaleate, and 1,4-naphthoquinone (NQ) exhibited complete protection against radiation-induced cell death in lymphocytes as estimated by propidium iodide staining. Radioprotection by NQ was demonstrated by inhibition of caspase activation, decrease in cell size, DNA fragmentation, nuclear blebbing, and clonogenic assay. Interestingly, NQ offered protection to lymphocytes even when added to cells postirradiation. NQ increased intracellular ROS levels and decreased GSH levels. NQ activated Nrf2 and increased the expression of the cytoprotective gene heme oxygenase-1 in lymphocytes. NQ increased ERK phosphorylation, which is upstream of Nrf2, and this ERK activation was through increased intracellular calcium levels. Administration of NQ to mice offered protection against whole-body irradiation (WBI)-induced apoptosis in splenic lymphocytes and loss of viability of spleen and bone marrow cells. It restored WBI-mediated changes in hematological parameters and functional responses of lymphocytes. Importantly, NQ rescued mice against WBI-induced mortality. These results demonstrated that a pro-oxidant such as NQ can protect against radiation-induced apoptosis by activation of multiple prosurvival mechanisms including activation of the calcium-ERK1/2-Nrf2 pathway.
Sigma-Aldrich
Diethyl maleate, ≥96%, FG
Sigma-Aldrich
Diethyl maleate, 97%