Merck
  • Chemistry of nitrosyliron complexes supported by a β-diketiminate ligand.
Chemistry of nitrosyliron complexes supported by a β-diketiminate ligand.
Inorganic chemistry (2011-01-20)
Zachary J Tonzetich, Florent Héroguel, Loi H Do, Stephen J Lippard
ABSTRACT
Several nitrosyl complexes of Fe and Co have been prepared using the sterically hindered Ar-nacnac ligand (Ar-nacnac = anion of [(2,6-diisopropylphenyl)NC(Me)](2)CH). The dinitrosyliron complexes [Fe(NO)(2)(Ar-nacnac)] (1) and (Bu(4)N)[Fe(NO)(2)(Ar-nacnac)] (2) react with [Fe(III)(TPP)Cl] (TPP = tetraphenylporphine dianion) to generate [Fe(II)(NO)(TPP)] and the corresponding mononitrosyliron complexes. The factors governing NO transfer with dinitrosyliron complexes (DNICs) 1 and 2 are evaluated, together with the chemistry of the related mononitrosyliron complex, [Fe(NO)Br(Ar-nacnac)] (4). The synthesis and properties of the related cobalt dinitrosyl [Co(NO)(2)(Ar-nacnac)] (3) is also discussed for comparison to DNICs 1 and 2. The solid-state structures of several of these compounds as determined by X-ray crystallography are reported.
Sigma-Aldrich
5,10,15,20-Tetraphenyl-21H,23H-porphine, ≥99%
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
5,10,15,20-Tetraphenyl-21H,23H-porphine, 97%