Merck
  • N-(3-methoxybenzyl)-(9Z,12Z,15Z)-octadecatrienamide promotes bone formation via the canonical Wnt/β-catenin signaling pathway.
N-(3-methoxybenzyl)-(9Z,12Z,15Z)-octadecatrienamide promotes bone formation via the canonical Wnt/β-catenin signaling pathway.
Phytotherapy research : PTR (2019-02-16)
Ting Wang, Chun-Han Sun, Hao-Bo Zhong, Yan Gong, Zhong-Kai Cui, Jing Xie, Yan-Peng Wang, Chuang Liang, He-He Cao, Xiao-Rui Chen, Zhi-Peng Zou, Sheng-Fa Li, Xiao-Chun Bai
ABSTRACT
Osteoporosis is characterized by low bone mineral density and microarchitectural deterioration of bone tissue. N-(3-methoxybenzyl)-(9Z,12Z,15Z)-octadecatrienamide (MBOC) is one of the macamides isolated from Maca (Lepidium meyenii Walp.), a cruciferous plant from the Andes of Peru. In this study, C3H/10T1/2 mesenchymal stem cells were treated with MBOC in osteogenic induction medium. An ovariectomized (OVX) mouse model was used to investigate the effect of 1-month MBOC treatment on the prevention of postmenopausal osteoporosis. Remarkably, trabecular thickness, trabecular number, and bone volume/tissue volume of the distal femoral metaphysis were significantly increased in OVX + MBOC mice compared with OVX mice, as revealed by microcomputed tomography analysis. Trabecular separation was decreased in OVX + MBOC mice compared with OVX mice. Consistently, MBOC increased the levels of osteocalcin and runt-related transcription factor 2 in OVX mice, as well as the expression of runt-related transcription factor 2, osterix, and alkaline phosphatase in C3H/10T1/2 cells. Mechanistically, MBOC activates the canonical Wnt/β-catenin signaling pathway via inhibiting phosphorylation of GSK-3β at Tyr216 and maintaining β-catenin expression. Collectively, the current study demonstrates the robustness of MBOC in the induction of mesenchymal stem cells osteogenic differentiation and consequent bone formation, suggesting that MBOC may be a potentially effective drug to treat postmenopausal osteoporosis.
Sigma-Aldrich
Anti-phospho-GSK3 (Tyr279/Tyr216) Antibody, clone 5G-2F, clone 5G-2F, Upstate®, from mouse
Sigma-Aldrich
Anti-SP7 antibody produced in rabbit, affinity isolated antibody