Merck
Fibrin structure and wound healing.
Journal of thrombosis and haemostasis : JTH (2006-05-13)
N Laurens, P Koolwijk, M P M de Maat
ABSTRACT
Fibrinogen and fibrin play an important role in blood clotting, fibrinolysis, cellular and matrix interactions, inflammation, wound healing, angiogenesis, and neoplasia. The contribution of fibrin(ogen) to these processes largely depends not only on the characteristics of the fibrin(ogen) itself, but also on interactions between specific-binding sites on fibrin(ogen), pro-enzymes, clotting factors, enzyme inhibitors, and cell receptors. In this review, the molecular and cellular biology of fibrin(ogen) is reviewed in the context of cutaneous wound repair. The outcome of wound healing depends largely on the fibrin structure, such as the thickness of the fibers, the number of branch points, the porosity, and the permeability. The binding of fibrin(ogen) to hemostasis proteins and platelets as well as to several different cells such as endothelial cells, smooth muscle cells, fibroblasts, leukocytes, and keratinocytes is indispensable during the process of wound repair. High-molecular-weight and low-molecular-weight fibrinogen, two naturally occurring variants of fibrin, are important determinants of angiogenesis and differ in their cell growth stimulation, clotting rate, and fibrin polymerization characteristics. Fibrin sealants have been investigated as matrices to promote wound healing. These sealants may also be an ideal delivery vehicle to deliver extra cells for the treatment of chronic wounds.
Sigma-Aldrich
TissueFab® bioink kit, (Gel)ma Fibrin (UV/365), low endotoxin
Sigma-Aldrich
TissueFab® bioink kit, (Gel)ma Fibrin (Vis/405), low endotoxin
Sigma-Aldrich
TissueFab® bioink , (GelAlgHA)MA Vis/405 nm, low endotoxin