Merck
  • Distribution of ACTH-like immunoreactivity in rat brain and gastrointestinal tract.
Distribution of ACTH-like immunoreactivity in rat brain and gastrointestinal tract.
Histochemistry (1978-04-04)
L I Larsson
PMID205525
ABSTRACT
Immunocytochemistry reveals ACTH-like immunoreactivity to reside not only in the pituitary but also in central nerves and in central nerves and in antral gastrin cells. In all probability, the central nerves store a peptide identical with or closely resembling true ACTH. Their pattern of distribution is, in some regions, similar to that of enkephalin-immunoreactive nerves. The antiserum demonstrating ACTH-like immunoreactivity in central nerves and in antral gastrin cells is directed towards the COOH-terminal part of the hormone. A peptide corresponding to this part, the corticotrophin-like intermediate peptide (CLIP) is manufactured by the pars intermedia of the pituitary. CLIP is devoid of adrenocortical activity, but has recently been shown to possess insulin-releasing activity. The occurrence of CLIP-like peptides in antral gastrin cells may indicate a role for such peptides in the gastrointestinal regulation of insulin release. The simultaneous occurrence of enkephalin-like and ACTH-like immunoreactivity in the antral gastrin cells is of particular interest since a large precursor molecule, containing both the enkephalin and the ACTH sequence has recently been identified.
Sigma-Aldrich
Adrenocorticotropic Hormone Fragment 18-39 human, ≥97% (HPLC)