Merck
  • ZAP-70 and SLP-76 regulate protein kinase C-theta and NF-kappa B activation in response to engagement of CD3 and CD28.
ZAP-70 and SLP-76 regulate protein kinase C-theta and NF-kappa B activation in response to engagement of CD3 and CD28.
Journal of immunology (Baltimore, Md. : 1950) (2001-04-21)
T M Herndon, X C Shan, G C Tsokos, R L Wange
ABSTRACT
The transcription factor NF-kappaB is a critical regulator of T cell function that becomes strongly activated in response to coengagement of TCR and CD28. Although events immediately proximal to NF-kappaB activation are well understood, uncertainty remains over which upstream signaling pathways engaged by TCR and CD28 lead to NF-kappaB activation. By using Jurkat T cell lines that are deficient or replete for either the protein tyrosine kinase ZAP-70 or the cytosolic adapter molecule SLP-76, the role of these proteins in modulating NF-kappaB activation was examined. NF-kappaB was not activated in response to coengagement of TCR and CD28 in either the ZAP-70- or SLP-76-negative cells, whereas stimuli that bypass these receptors (PMA plus A23187, or TNF-alpha) activated NF-kappaB normally. Protein kinase C (PKC) theta activation, which is required for NF-kappaB activation, also was defective in these cells. Reexpression of ZAP-70 restored PKCtheta and NF-kappaB activation in response to TCR and CD28 coengagement. p95(vav) (Vav)-1 tyrosine phosphorylation was largely unperturbed in the ZAP-70-negative cells; however, receptor-stimulated SLP-76/Vav-1 coassociation was greatly reduced. Wild-type SLP-76 fully restored PKCtheta and NF-kappaB activation in the SLP-76-negative cells, whereas 3YF-SLP-76, which lacks the sites of tyrosine phosphorylation required for Vav-1 binding, only partially rescued signaling. These data illustrate the importance of the ZAP-70/SLP-76 signaling pathway in CD3/CD28-stimulated activation of PKC theta and NF-kappaB, and suggest that Vav-1 association with SLP-76 may be important in this pathway.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Anti-PECAM-1 Antibody, domains 3-6 of human PECAM-1, clone HC1/6, clone HC1/6, Chemicon®, from mouse