Merck
  • Synthesis of novel 3-amino and 29-hydroxamic acid derivatives of glycyrrhetinic acid as selective 11β-hydroxysteroid dehydrogenase 2 inhibitors.
Synthesis of novel 3-amino and 29-hydroxamic acid derivatives of glycyrrhetinic acid as selective 11β-hydroxysteroid dehydrogenase 2 inhibitors.
Bioorganic & medicinal chemistry (2010-09-21)
Christian Stanetty, Laszlo Czollner, Iris Koller, Priti Shah, Rawindra Gaware, Thierry Da Cunha, Alex Odermatt, Ulrich Jordis, Paul Kosma, Dirk Classen-Houben
ABSTRACT
Glycyrrhetinic acid, the metabolite of the natural product glycyrrhizin, is a well known nonselective inhibitor of 11β-hydroxysteroid dehydrogenase (11β-HSD) type 1 and type 2. Whereas inhibition of 11β-HSD1 is currently under consideration for treatment of metabolic diseases, such as obesity and diabetes, 11β-HSD2 inhibitors may find therapeutic applications in chronic inflammatory diseases and certain forms of cancer. So far, no selective 11β-HSD2 inhibitor has been developed and neither animal studies nor clinical trials have been reported based on 11β-HSD2 inhibition. Starting from the lead compound glycyrrhetinic acid, novel triterpene type derivatives were synthesized and analyzed for their biological activity against overexpressed human 11β-HSD1 and 11β-HSD2 in cell lysates. Several hydroxamic acid derivatives showed high selectivity for 11β-HSD2. The most potent and selective compound is active against human 11β-HSD2 in the low nanomolar range with a 350-fold selectivity over human 11β-HSD1.
Sigma-Aldrich
18β-Glycyrrhetinic acid, 97%