Merck
  • TROP-2 immunohistochemistry: a highly accurate method in the differential diagnosis of papillary thyroid carcinoma.
TROP-2 immunohistochemistry: a highly accurate method in the differential diagnosis of papillary thyroid carcinoma.
Pathology (2016-06-18)
Andrey Bychkov, Pichet Sampatanukul, Shanop Shuangshoti, Somboon Keelawat
ABSTRACT
We aimed to evaluate the diagnostic utility of the novel immunohistochemical marker TROP-2 on thyroid specimens (226 tumours and 207 controls). Whole slide immunohistochemistry was performed and scored by automated digital image analysis. Non-neoplastic thyroid, follicular adenomas, follicular carcinomas, and medullary carcinomas were negative for TROP-2 immunostaining. The majority of papillary thyroid carcinoma (PTC) specimens (94/114, 82.5%) were positive for TROP-2; however, the pattern of staining differed significantly between the histopathological variants. All papillary microcarcinomas (mPTC), PTC classic variant (PTC cv), and tall cell variant (PTC tcv) were TROP-2 positive, with mainly diffuse staining. In contrast, less than half of the PTC follicular variant specimens were positive for TROP-2, with only focal immunoreactivity. TROP-2 could identify PTC cv with 98.1% sensitivity and 97.5% specificity. ROC curve analysis found that the presence of >10% of TROP-2 positive cells in a tumour supported a diagnosis of PTC. The study of intratumoural heterogeneity showed that low-volume cytological samples of PTC cv could be adequately assessed by TROP-2 immunostaining. The TROP-2 H-score (intensity multiplied by proportion) was significantly associated with PTC variant and capsular invasion in encapsulated PTC follicular variant (p<0.001). None of the baseline (age, gender) and clinical (tumour size, nodal disease, stage) parameters were correlated with TROP-2 expression. In conclusion, TROP-2 membranous staining is a very sensitive and specific marker for PTC cv, PTC tcv, and mPTC, with high overall specificity for PTC.
Sigma-Aldrich
TROP2 (EP431) Rabbit Monoclonal Primary Antibody