Merck
  • Possible therapeutic applicability of galectin-9 in cutaneous T-cell lymphoma.
Possible therapeutic applicability of galectin-9 in cutaneous T-cell lymphoma.
Journal of dermatological science (2019-12-04)
Rina Nakajima, Tomomitsu Miyagaki, Hiroaki Kamijo, Tomonori Oka, Naomi Shishido-Takahashi, Hiraku Suga, Makoto Sugaya, Shinichi Sato
ABSTRACT
Galectin-9, a member of the galectin family, can promote tumor growth through inducing apoptosis in anti-tumor immune cells via T cell immunoglobulin and mucin domain 3 (TIM-3). On the other hand, galectin-9 also induces tumor cell apoptosis in many malignancies and thought to have potential as an anti-cancer agent. To examine the expression and therapeutic applicability of galectin-9 in cutaneous T-cell lymphoma (CTCL). Galectin-9 expression in lesional skin and sera was measured using CTCL samples. The effect of galectin-9 on CTCL cell lines was investigated in vitro. We also examined effect of galectin-9 on tumor growth of CTCL cells in immune-deficient mice. Moreover, we examined the efficacy of galectin-9, anti-TIM-3 blocking antibody, or their combination on tumor growth of EL-4 cells in wild-type mice. Galectin-9 was expressed on tumor cells in lesional skin of CTCL and the expression levels were associated with decreased CD8+ T-cell infiltration. Serum galectin-9 levels were correlated with disease severity markers. High-dose galectin-9 induced cell death of CTCL cell lines through activation of caspase-3 and caspase-9, independently of TIM-3. High-dose galectin-9 suppressed the growth of CTCL cells and EL-4 cells in vivo. Furthermore, additional anti-TIM-3 blocking antibody administration to galectin-9 achieved greater inhibition of tumor growth compared to single administration. Galectin-9 expression on tumor cells may be associated with CTCL progression through attenuating anti-tumor immunity. On the other hand, exogenous high-dose galectin-9 administration can be a therapeutic strategy for CTCL and anti-TIM-3 blocking antibody can augment the efficacy of galectin-9.
Sigma-Aldrich
Galectin-9 human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)