Merck
  • Hypoxia-inducible factor-1 signaling pathway influences the sensitivity of HCC827 cells to gefitinib.
Hypoxia-inducible factor-1 signaling pathway influences the sensitivity of HCC827 cells to gefitinib.
Oncology letters (2019-03-19)
Qian Jin, Jianying Zhou, Xianrong Xu, Feihua Huang, Weihua Xu
ABSTRACT
The majority of patients with non-small cell lung cancer (NSCLC) with activating epidermal growth factor receptor (EGFR) mutations inevitably progress in stage despite an initial substantial and rapid response to EGFR-tyrosine kinase inhibitors (EGFR-TKIs). Previous research indicates that hypoxia may be associated with resistance to EGFR-TKIs in EGFR mutation-positive NSCLC. Therefore, the present study regulated the activity of hypoxia-inducible factor-1 (HIF-1) signaling pathway to observe if it is able to alter the sensitivity of lung cancer cells to gefitinib. The present study selected 3-(5'-hydroxymethyl-2'-furyl)-1-benzylindazole (YC-1) and dimethyloxalylglycine (DMOG) as a HIF-1 signaling pathway inhibitor and activator, respectively, on HCC827 cells. Cells were incubated with different treatments for different durations: A blank control, DMOG, gefitinib, or DMOG and gefitinib combined, for 36 and 48 h; and then a blank control, YC-1, gefitinib, or YC-1 and gefitinib combined, for 16 and 28 h. A western blot analysis assay was performed to evaluate the protein expression levels of HIF-1α and phosphorylated hepatocyte growth factor receptor (p-MET), an MTT assay was used to determine cell proliferation, a colony formation assay was used to investigate the colony-forming ability and a wound healing assay was used to test the cell migration ability. Additionally, Pearson's correlation analysis was used to evaluate the correlation between p-Met and HIF-1α expression levels. Finally, it was identified that gefitinib and DMOG combined notably improve the growth and cell migration ability of HCC827 cells, compared with gefitinib alone. When gefitinib and YC-1 were combined, the inhibiting effect on the growth and cell migration ability of HCC827 cells was substantially enhanced, compared with the control cells. Pearson's correlation analysis revealed that the p-Met expression level had a strong positive correlation with HIF-1α expression levels. Thus, it was concluded that the HIF-1 signaling pathway influences the sensitivity of HCC827 cells to gefitinib. The positive correlation between p-Met and HIF-1α expression levels may be the underlying mechanism of the HIF-1 signaling pathway influencing the sensitivity of HCC827 cells to gefitinib.
Sigma-Aldrich
HRP-conjugated Anti-Rabbit IgG Concentrate (Item I1)