Merck
  • Decreased placental and muscular expression of the fibroblast growth factor 19 in gestational diabetes mellitus.
Decreased placental and muscular expression of the fibroblast growth factor 19 in gestational diabetes mellitus.
Journal of diabetes investigation (2018-05-08)
Dongyu Wang, Shuqia Xu, Wenjing Ding, Caixia Zhu, Songqing Deng, Xiwen Qiu, Zilian Wang
ABSTRACT
Fibroblast growth factor (FGF)19 has been shown to improve glycemic homeostasis and lipid metabolism in animal models. In humans, decreased FGF19 level has been described in diabetes. The present study aimed to investigate the expression of FGF19 in gestational diabetes mellitus (GDM) patients. Samples for measurement were obtained from 20 women with GDM and 25 healthy controls. The messenger ribonucleic acid (mRNA) and protein expression levels of FGF19, FGF21 and co-receptor β-klotho (KLB) in the placenta, rectus muscle and subcutaneous fat tissues were quantified by real-time quantitative polymerase chain reaction, western blot and immunohistochemistry, respectively. Women with GDM had significantly lower mRNA and protein expressions of FGF19 than control women in the placenta (mRNA 0.33 ± 0.05 vs 0.72 ± 0.09; protein 0.34 ± 0.13 vs 0.85 ± 0.20) and rectus muscle (mRNA 0.83 ± 0.11 vs 1.28 ± 0.19; protein 0.78 ± 0.24 vs 1.23 ± 0.39). However, there were no significant differences between GDM women and controls with respect to the expression levels of FGF21 and β-klotho in the placenta and rectus muscle. There were almost no detectable FGF19 and FGF21 expressions in subcutaneous fat tissue. Furthermore, β-klotho expression levels were not different between the GDM and control group in subcutaneous fat. FGF19 expressions are decreased in the placenta and rectus muscle of women with GDM. This might contribute to the pathophysiology or development of GDM.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal Anti-FGF21 antibody produced in mouse, clone 2F11, purified immunoglobulin, buffered aqueous solution