
Resorufin ethyl ether

Synonym(s):
7-Ethoxy-3H-phenoxazin-3-one, Ethoxyresorufin, O7-Ethylresorufin
Empirical Formula (Hill Notation):
C14H11NO3
CAS Number:
Molecular Weight:
241.24
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing