
Iopromide for system suitability 1

Synonym(s):
Iopromide
Empirical Formula (Hill Notation):
C18H24I3N3O8
CAS Number:
Molecular Weight:
791.11
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing