
Ibandronate sodium monohydrate

Synonym(s):
Ibandronate monosodium salt monohydrate
Empirical Formula (Hill Notation):
C9H22NNaO7P2 · H2O
CAS Number:
Molecular Weight:
359.23
Products
Formula Weight
Manufacturer
API Family
Showing 1-1 of 1
Compare
Product Number
Description
Pricing