
Buflomedil hydrochloride

Synonym(s):
4-(1-Pyrrolidinyl)-1-(2,4,6-trimethoxyphenyl)-1-butanone
Empirical Formula (Hill Notation):
C17H25NO4 · HCl
CAS Number:
Molecular Weight:
343.85
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing