Skip to Content
Merck
All Photos(1)

Key Documents

5.31057

Sigma-Aldrich

ETV1 Inhibitor, BRD32048

Synonym(s):

ETV1 Inhibitor, BRD32048, N-(4-Methoxyphenyl)-6-(1-piperidinylmethyl)-1,3,5-triazine-2,4-diamine, ER81 Inhibitor, ETV1-mediated Transcription Inhibitor, Transcription Inhibitor ETV1 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22N6O
Molecular Weight:
314.39
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable triazine compound that directly and reversibly binds to ETS family transcription factor ETV1 (KD = 17.1 µM) and reduces its transcriptional activity, independent of its binding to DNA. Selectively inhibits p300-dependent ETV1 acetylation at K33 and K116 and thereby promotes its degradation (~50 µM). Inhibits the invasion capacity of ETV1-dependent cell lines and primary melanocytes expressing NRASG12D, but not in primary melanocytes expressing NRAS. Does not affect PC3 cell line that is not dependent on ETV1 for invasion. Also it has no effect on the invasion of cancer cells expressing mutated ETV1 (K33R/K116R).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
A cell-permeable triazine compound that targets the ETS family transcription factor ETV1, but not TBX21, RELA, RUNX1, RFWD2, or P/CAF, via affinity interaction (KD = kd/ka = 5.853 x 10-2 s-1/4.128 x 103 M-1s-1= 14.2 µM) and selectively suppresses ETV1-, but not MITF-, dependent reporter transcription activity in 501mel melanoma cells (2.57-fold vs 4-fold of mock level in 24 h, respectively, with or without 10 µM BRD32048) without affecting ETV1 DNA-binding activity. Shown to inhibit ETV1-dependent invasion by downregulating p300-, but not P/CAF-, mediated ETV1 lysine acetylation (K-Ac) in LNCaP & 501mel cultures (50 µM for 16 h) that are known to harbor ETV1 chromosomal rearrangement (translocation) and amplification, respectively, resulting in enhanced ETV1 degradation, while ETV1 K-Ac is not detectable in PC3 (lacking ETV1 rearrangement) & SK-MEL-28 (wild-type ETV1) cells and no invasion inhibition by BRD32048 treatment is seen in these cultures. Likewise, BRD32048 is demonstrated to inhibit invasion of NRASG12D-expressing primary melanocytes (pmel) only when infected to express ETV1, and no inhibition is seen in the non-infected or MYC-infected cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ETS family transcription factor ETV1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Pop, M.S., et al. 2014. Mol. Cancer Ther.13, 1492.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service