Merck
HPLC Analysis of Heroin, Cocaine, Amphetamine and MDMA on Ascentis® Express HILIC
HPLC Analysis of Heroin, Cocaine, Amphetamine and MDMA on Ascentis® Express HILIC application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express HILIC, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
standard
Product No.
Description
Pricing
D-Amphetamine hemisulfate salt solution
1.0 mg/mL in methanol, analytical standard, for drug analysis
(±)-3,4-Methylenedioxymethamphetamine hydrochloride solution
analytical standard, for drug analysis
Heroin
analytical standard, ≥98% (HPLC)
CONDITIONS
column
Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 μm particles (53939-U)
mobile phase
[A] 10 mM ammonium acetate in water, pH 3.5; [B] 10 mM ammonium acetate in acetonitrile; (2.98, A:B)
flow rate
0.4 mL/min
column temp.
50 °C
detector
ESI(+), m/z 100-400
injection
1 μL
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany