HPLC Analysis of Amphetine, Methamphetamine, MDA, and MDMA on Discovery® HS F5
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery HS F5, 15 cm × 4.6 mm I.D., 5 μm particles (567516-U)
mobile phase
[A] 10 mM ammonium acetate in [B] water: acetonitrile (10:90)
flow rate
1.5 mL/min
column temp.
35 °C
detector
UV, 210 nm
injection
5 μL
sample
50 μg/mL each (amphetamine, methamphetamine, MDA and MDMA) in methanol
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany