US EPA Method 608.1/608.2: GC Analysis of Organochlorine Pesticides on SPB®-608
Materials
analytical column
Product No.
Description
Pricing
accessory
Product No.
Description
Pricing
Wheaton wide-mouth bottle
amber soda-lime glass bottle, capacity 1000 mL, white polypropylene cap, (PTFE faced LDPE liner), case of 12 eaguard column
recommended
Product No.
Description
Pricing
CONDITIONS
column
SPB-608, 30 m x 0.25 mm I.D., 0.25 μm (24103-U)
oven
75 °C (2 min), 15 °C/min to 300 °C (5 min)
inj. temp.
250 °C
detector
ECD, 300 °C
carrier gas
helium, 1.2 mL/min. constant
injection
1 μL, splitless (splitter open after 0.75 min)
liner
4 mm I.D., single taper
sample
mixture of 608.1 and 608.2 calibration solutions (40351-U and 40352-U) diluted to 100 ppb in n-hexane
Description
Legal Information
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany