Merck
NMR of molecular endofullerenes dissolved in a nematic liquid crystal.
Physical chemistry chemical physics : PCCP (2017-04-27)
Karel Kouřil, Christopher Wickens, Benno Meier, Shamim Alom, John Gräsvik, Richard J Whitby, Malcolm H Levitt
ABSTRACT
We report the NMR of the molecular endofullerenes H
Sigma-Aldrich
N-(4-Methoxybenzylidene)-4-butylaniline, 98%