Merck
  • Differential viral haemorrhagic septicaemia virus genotype IVb infection in fin fibroblast and epithelial cell lines from walleye, Sander vitreus (Mitchill), at cold temperatures.
Differential viral haemorrhagic septicaemia virus genotype IVb infection in fin fibroblast and epithelial cell lines from walleye, Sander vitreus (Mitchill), at cold temperatures.
Journal of fish diseases (2015-02-04)
N T K Vo, A W Bender, J S Lumsden, B Dixon, N C Bols
ABSTRACT
A cell line, WE-cfin11e, with an epithelial-like morphology was developed from a caudal fin of walleye, Sander vitreus (Mitchill), characterized as distinct from the established walleye caudal fin fibroblast-like cell line, WE-cfin11f, and compared with WE-cfin11f for susceptibility to VHSV IVb. Immunocytochemistry and confocal microscopy were used to localize the intermediate filament protein, vimentin, the tight junction protein, zonula occludens-1 (ZO-1), the extracellular matrix protein, collagen I, and the viral protein, G. Although both cell lines contained vimentin, only WE-cfin11e stained for ZO-1 and only WE-cfin11f stained for collagen I. Ascorbic acid increased the accumulation of collagen I and caused the appearance of collagen fibres only in WE-cfin11f cultures. At 14 °C, both cell lines produced VHSV IVb, but the infection developed more rapidly in WE-cfin11f. At 4 °C, both cell lines became infected with VHSV IVb as judged by the expression of viral proteins, N and G, but only WE-cfin11f produced virus. The results suggest that cold temperatures can modulate viral tropism.
Sigma-Aldrich
MISSION® esiRNA, targeting human TJP1
Sigma-Aldrich
Triton X-100, laboratory grade