Merck
  • Effects of Intraduodenal Infusions of L-phenylalanine and L-glutamine on Antropyloroduodenal Motility and Plasma Cholecystokinin in Healthy Men.
Effects of Intraduodenal Infusions of L-phenylalanine and L-glutamine on Antropyloroduodenal Motility and Plasma Cholecystokinin in Healthy Men.
Journal of neurogastroenterology and motility (2015-07-02)
Robert E Steinert, Maria F Landrock, Michael Horowitz, Christine Feinle-Bisset
ABSTRACT
Dietary proteins have potent eating-inhibitory and glucose-lowering effects, which may be mediated via effects of amino acids on gastrointestinal hormone and motor function, although little information is available. We have now evaluated the effects of L-phenylalanine (L-Phe) and L-glutamine (L-Gln) on antropyloroduodenal motility and plasma cholecystokinin (CCK) concen-trations. Two double-blind, 3-way cross-over studies were performed, each including 10 healthy, normal-weight men. We determined the antropyloroduodenal motor and plasma CCK responses to 90-minute intraduodenal infusions of L-Phe (study A) or L-Gln (study B), each at 0.15 kcal/min (total 13.5 kcal), or 0.45 kcal/min (total 40.5 kcal), or saline (control), in randomized fashion. Intraduodenal L-Phe at 0.45 kcal/min, but not at 0.15 kcal/min, suppressed antral (P < 0.01), and stimulated phasic (P < 0.01), but not tonic, pyloric, or duodenal pressures, while L-Phe at both 0.15 kcal/min and 0.45 kcal/min stimulated plasma CCK. In contrast, L-Gln had no effect on antral, duodenal or pyloric pressures, or plasma CCK. Intraduodenal infusions of L-Phe and L-Gln, in doses of 0.15 kcal/min and 0.45 kcal/min for 90 minutes, have different effects on antropyloroduodenal motility and CCK in normal-weight men. The modulation of antral and pyloric pressures and CCK may contribute to the eating-inhibitory effects of oral L-Phe, possibly through the slowing of gastric emptying.
Sigma-Aldrich
Anti-Cholecystokinin (26-33) (CCK-8) antibody produced in rabbit, whole antiserum
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder