Merck
  • PpPIF-1: first isolated full-length PIF-like element from the bamboo Phyllostachys pubescens.
PpPIF-1: first isolated full-length PIF-like element from the bamboo Phyllostachys pubescens.
Genetics and molecular research : GMR (2012-05-12)
M B Zhou, X M Liu, D Q Tang
ABSTRACT
PIF-like elements are the first-described members of a recently discovered and widespread superfamily of DNA transposons, named PIF/Harbinger. Complete and partial PIF-like elements have been isolated from hundreds of plant species. Previously, we identified 139 partial PIF-like transposases in the Bambusoideae, of which three were from the bamboo species Phyllostachys pubescens. Here we report identification and isolation of the first full-length PIF-like element (PpPIF-1) from P. pubescens; identification was made by chromosome walking, based on a modified magnetic enrichment procedure that allows efficient cloning of flanking sequences up to 3 kb in length. PpPIF-1 is 5953 bp in length, with 20-bp imperfect inverted terminal repeats and 3-bp target site duplications. This element contains two open reading frames, one encoding a putative transposase, including the complete DDE-domain typical of PIF/Harbinger elements from plants, and the other encoding a DNA-binding protein. There are seven termination codons and two frameshift mutations in the open reading frames, probably due to vertical inactivation.
Product Description
ECL Direct Nucleic Acid, Cytiva RPN3000, pack of 1 ea
ECL Detection Reagents, Cytiva RPN3004, pack of 1 ea
ECL Direct Nucleic Acid, Cytiva RPN3005, pack of 1 ea
ECL Direct Nucleic Acid, Cytiva RPN3001, pack of 1 ea