Merck
  • Chlorogenic acid attenuates ventricular remodeling after myocardial infarction in mice.
Chlorogenic acid attenuates ventricular remodeling after myocardial infarction in mice.
International heart journal (2013-06-19)
Yoshinori Kanno, Ryo Watanabe, Hirofumi Zempo, Masahito Ogawa, Jun-Ichi Suzuki, Mitsuaki Isobe
ABSTRACT
Chlorogenic acid (CGA), which is a key component of coffee, has many biological effects such as anti-inflammation activity. However, the effects of CGA on ventricular remodeling after myocardial ischemia have not been well investigated. To test the hypothesis that CGA can attenuate chronic ventricular remodeling after myocardial ischemia, we orally administered CGA to murine myocardial ischemia models. Seven to nine week-old C57BL/6 mice were used. A myocardial infarction (MI) model was produced by permanent ligation of the left anterior descending coronary artery (LAD) using an 8-0 suture passed under the arteries. These mice were randomly assigned into 4 groups in each experimental model. Some MI mice were supplemented orally with CGA (30 mg/kg/day, MI+CGA group, n = 13) as a CGAtreated MI group, and other MI mice received vehicle (MI+vehicle group, n = 11) as a vehicle-treated MI group. Shamoperated mice without MI also received vehicle (Sham+vehicle group, n = 3) as a sham group, and sham-operated mice without MI received CGA (30 mg/kg/day, Sham+CGA group, n = 8) as a Sham+CGA group. Just before sacrifice on day 14, we measured blood pressure and heart rate and performed echocardiography. We obtained 3 transverse sections per heart for histopathologic examination. There were no differences in body weight, heart rate, or blood pressure among the groups on day 14. The vehicle-treated MI group showed significantly impaired left ventricular contraction compared to the sham-operated group. However, the CGA-treated MI group showed significantly improved ventricular contraction compared to the vehicle-treated MI group. Severe myocardial fibrosis with enhanced macrophage infiltration was observed in the vehicle-treated ischemia group on day 14. CGA attenuated these fibrotic changes with suppressed macrophage infiltration without systemic adverse effects. CGA may effectively suppress chronic ventricular remodeling after myocardial ischemia because it is critically involved in the suppression of macrophage infiltration.
Product Description
Chlorogenic acid, primary reference standard
Pricing and availability is not currently available.
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Pricing and availability is not currently available.