Merck
  • Highly hydrophobic electrospun fiber mats from polyisobutylene-based thermoplastic elastomers.
Highly hydrophobic electrospun fiber mats from polyisobutylene-based thermoplastic elastomers.
Biomacromolecules (2011-04-01)
Goy Teck Lim, Judit E Puskas, Darrell H Reneker, Antal Jákli, Walter E Horton
ABSTRACT
This paper is the first report of electrospinning neat polyisobutylene-based thermoplastic elastomers. Two generations of these materials are investigated: a linear poly(styrene-b-isobutylene-b-styrene) (L_SIBS) triblock copolymer and a dendritic poly(isobutylene-b-p-methylstyrene) (D_IB-MS), also a candidate for biomedical applications. Cross-polarized optical microscopy shows birefringence, indicating orientation in the electrospun fibers, which undergo large elongation and shear during electrospinning. In contrast to the circular cross section of L_SIBS fibers, D_IB-MS yields dumbbell-shaped fiber cross sections for the combination of processing conditions, molecular weight, and architecture. Hydrophobic surfaces with a water contact angle as high as 146 ± 3° were obtained with D_IB-MS that had the noncircular fiber cross section and a hierarchical arrangement of nano- to micrometer-sized fibers in the mat. These highly water repellent fiber mats were found to serve as an excellent scaffold for bovine chondrocytes to produce cartilage tissue.
Sigma-Aldrich
Polyisobutylene, average Mw ~1,000,000, average Mn ~600,000 by GPC/MALLS, average Mv ~1,200,000
Sigma-Aldrich
Polyisobutylene, average Mw ~500,000, average Mn ~200,000 by GPC/MALLS, average Mv ~420,000