Merck
  • Effect of co-initiator ratio on the polymer properties of experimental resin composites formulated with camphorquinone and phenyl-propanedione.
Effect of co-initiator ratio on the polymer properties of experimental resin composites formulated with camphorquinone and phenyl-propanedione.
Dental materials : official publication of the Academy of Dental Materials (2008-10-14)
Luis Felipe J Schneider, Larissa M Cavalcante, Simonides Consani, Jack L Ferracane
ABSTRACT
To evaluate the effect of amine ratio (ethyl 4-dimethylaminobenzoate, EDMAB) on the maximum rate of polymerization (R(p)(max)), degree of conversion (DC), Knoop hardness (KH), water sorption (Wsp), water solubility (Wsl) and color changes (DeltaE) over time of resin composites formulated with the photoinitiators camphorquinone (CQ), phenylpropanedione (PPD) and CQ-PPD in combination. Experimental resin composites were made with photoinitiator:amine ratios of 2:1, 1:1, 1:1.5 and 1:2 by weight. R(p)(max) was evaluated with differential scanning calorimetry (DSC), DC with DSC and Fourier transformed infrared (FTIR) spectroscopy, KH with Knoop indentation, Wsp and Wsl adapted from ISO 4049; and color with a chromameter. The results were analyzed with two-way ANOVA/Tukey's multiple comparison test (p<0.05). The higher the amine ratio in the composite, the higher was DC, R(p)(max), and KH, and the lower was Wsl, regardless of the photoinitiator type. The use of PPD alone resulted in poorer properties than CQ and CQ-PPD. Many factors seem to affect the color changes and the b-axis data revealed that the higher the amine ratio, the higher was the +b value (yellowing) for CQ and CQ-PPD formulations. Higher amine ratios led to improved polymer properties, but also produced more yellowing in resin composites with CQ and CQ-PPD. The use of PPD alone was not advantageous for producing good final properties when compared to CQ and CQ-PPD.
Sigma-Aldrich
1-Phenyl-1,2-propanedione, 98%, FG
Sigma-Aldrich
1-Phenyl-1,2-propanedione, 99%