Merck
  • Loss of Family with Sequence Similarity 13, Member A Exacerbates Pulmonary Fibrosis Potentially by Promoting Epithelial to Mesenchymal Transition.
Loss of Family with Sequence Similarity 13, Member A Exacerbates Pulmonary Fibrosis Potentially by Promoting Epithelial to Mesenchymal Transition.
The Kobe journal of medical sciences (2020-02-08)
Elda Putri Rahardini, Koji Ikeda, Dhite Bayu Nugroho, Ken-Ichi Hirata, Noriaki Emoto
ABSTRACT
Idiopathic pulmonary fibrosis (IPF) is a devastating disease with poor prognosis due to limited clinical treatment options. IPF is characterized by the augmented deposition of extracellular matrix driven by myofibroblasts, and the epithelial-mesenchymal transition (EMT) has been known to play an essential role in the mechanism of pulmonary fibrosis. Previous genome-wide association study identified Fam13a as one of genes that showed genetic link with IPF and chronic obstructive pulmonary disease. Here, we analyzed the role of Fam13a in the pathogenesis of pulmonary fibrosis using Fam13a-deficient mice. We found that Fam13a was down-regulated in mouse lungs of bleomycin-induced pulmonary fibrosis model. Of note, genetic deletion of Fam13a exacerbated the lung fibrosis induced by bleomycin in association with enhanced EMT in mice. Moreover, silencing of Fam13a accelerated EMT induced by TGF-β and TNF-α in alveolar epithelial cells, accompanied by increased active β-catenin and its nuclear accumulation. Our data revealed a crucial role of Fam13a in the development of pulmonary fibrosis potentially through inhibiting EMT, and thus Fam13a has a therapeutic potential in the treatment of IPF.
Sigma-Aldrich
Anti-FAM13A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - FITC antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture